Thermo Scientific Chemicals

p-Toluenesulfonylmethyl isocyanide, 97%, Thermo Scientific Chemicals

Catalog number: A14312.06
5 g, Each
Thermo Scientific Chemicals

p-Toluenesulfonylmethyl isocyanide, 97%, Thermo Scientific Chemicals

Catalog number: A14312.06
5 g, Each
Quantity
Catalog number: A14312.06
also known as A14312-06
Price (USD)
Price: 38.00
Online price: 36.65
Your price:
Quantity
-

Chemical Identifiers

CAS
36635-61-7
IUPAC Name
1-isocyanomethanesulfonyl-4-methylbenzene
Molecular Formula
C9H9NO2S
InChI Key
CFOAUYCPAUGDFF-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)S(=O)(=O)C[N+]#[C-]
Appearance (Color)
White to cream or pale yellow to pale brown
Clarity
(2.5% soln. in Acetone): Clear to almost clear
Identification (FTIR)
Conforms
Assay (HPLC)
≥96.0%
Form
Crystalline powder or powder

Description

p-Toluenesulfonylmethyl isocyanide is a synthetic reagent used in the synthesis of triplet drugs with the 1,3,5-trioxazatriquinane skeleton, pyrroles, oxazoles and in imidazoles. It serves as an isonitrile component in a diastereoselective Passerini reaction. Furthermore, it is used in the Van Leusen reaction to convert aldehydes to nitriles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonylmethyl isocyanide is a synthetic reagent used in the synthesis of triplet drugs with the 1,3,5-trioxazatriquinane skeleton, pyrroles, oxazoles and in imidazoles. It serves as an isonitrile component in a diastereoselective Passerini reaction. Furthermore, it is used in the Van Leusen reaction to convert aldehydes to nitriles.

Solubility
Soluble in chloroform and methanol. Insoluble in water.

Notes
Moisture, air and light sensitive. Store in cool place. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text