Thermo Scientific Chemicals

2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, 98+ atom % D, Thermo Scientific Chemicals

Catalog number: A14489.MD
250 mg, Each
Thermo Scientific Chemicals

2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, 98+ atom % D, Thermo Scientific Chemicals

Catalog number: A14489.MD
250 mg, Each
Quantity
Catalog number: A14489.MD
also known as A14489-MD
Price (USD)

Chemical Identifiers

CAS
24493-21-8
IUPAC Name
sodium 3-(trimethylsilyl)(²H₄)propanoate
Molecular Formula
C6H13NaO2Si
InChI Key
OIIWPAYIXDCDNL-HGFPCDIYSA-M
SMILES
[Na+].[2H]C([2H])(C([O-])=O)C([2H])([2H])[Si](C)(C)C
Form
Powder
Elemental Analysis
≥98.0 atom% deuterium
Water Content (Karl Fischer Titration)
≤2.0%
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥92.5% to ≤107.5%

Description

2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.

Solubility
Soluble in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text