Thermo Scientific Chemicals

1,1,1-Trifluoro-2,4-pentanedione, 98%, Thermo Scientific Chemicals

Catalog number: A14682.06
5 g, Each
Thermo Scientific Chemicals

1,1,1-Trifluoro-2,4-pentanedione, 98%, Thermo Scientific Chemicals

Catalog number: A14682.06
5 g, Each
Quantity
Catalog number: A14682.06
also known as A14682-06
Price (USD)
Price: 29.80
Online price: 25.65
Your price:
Quantity
-

Chemical Identifiers

CAS
367-57-7
Form
Liquid
Refractive Index
1.3855-1.3900 @ 20?C
Appearance (Color)
Clear colorless to yellow
Comment
Material sourced in UK and US
Assay (GC)
≥97.5% (sum of tautomers)

Description

1,1,1-Trifluoro-2,4-pentanedione has been used as reagent in the preparation of 2-alkylcarbonyl and 2-benzoyl-3-trifluoromethylquinoxaline 1,4-di-N-oxide derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Trifluoro-2,4-pentanedione has been used as reagent in the preparation of 2-alkylcarbonyl and 2-benzoyl-3-trifluoromethylquinoxaline 1,4-di-N-oxide derivatives.

Solubility
Soluble in water (slightly) and many organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text