Thermo Scientific Chemicals

1,3-Dibromopropane, 98%, Thermo Scientific Chemicals

Catalog number: A14687.0B
1000 g, Each
Thermo Scientific Chemicals

1,3-Dibromopropane, 98%, Thermo Scientific Chemicals

Catalog number: A14687.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A14687.0B
also known as A14687-0B
Price (USD)
Price: 208.00
Online price: 176.65
Your price:
Quantity
-

Chemical Identifiers

CAS
109-64-8
IUPAC Name
1,3-dibromopropane
Molecular Formula
C3H6Br2
InChI Key
VEFLKXRACNJHOV-UHFFFAOYSA-N
SMILES
BrCCCBr
Form
Liquid
Refractive Index
1.5210-1.5250 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms

Description

1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text