Thermo Scientific Chemicals

4-Methylbiphenyl, 98%, Thermo Scientific Chemicals

Catalog number: A14702.06
5 g, Each
Thermo Scientific Chemicals

4-Methylbiphenyl, 98%, Thermo Scientific Chemicals

Catalog number: A14702.06
5 g, Each
Quantity
Catalog number: A14702.06
also known as A14702-06
Price (USD)
Price: 120.00
Online price: 103.65
Your price:
Quantity
-

Chemical Identifiers

CAS
644-08-6
Assay (GC)
≥97.5% (U.K. specification)
Melting Point (clear melt)
43.5-49.5°C (U.K. specification)
Appearance (Color)
White to cream or pale yellow to pale brown
Assay from Supplier's CofA
≥97.5% (GC) (U.S. specification)
Form
Crystals or powder or crystalline powder or fused/lumpy solid

Description

4-Methylbiphenyl, is used as a flavoring ingredient in food industry. It is also used as an intermediate in agrochemcial, dyestuff, flavor and fragrance, pharmaceutical, syntheses material. It is also used as an OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbiphenyl, is used as a flavoring ingredient in food industry. It is also used as an intermediate in agrochemcial, dyestuff, flavor and fragrance, pharmaceutical, syntheses material. It is also used as an OLED intermediate.

Solubility
Insoluble in water. Soluble in alcohol and ether, very soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text