Thermo Scientific Chemicals

4-Nitrobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14738.30
250 g, Each
Thermo Scientific Chemicals

4-Nitrobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14738.30
250 g, Each
Quantity
Catalog number: A14738.30
also known as A14738-30
Price (USD)
Price: 26.20
Online price: 22.65
Your price:
Quantity
-

Chemical Identifiers

CAS
62-23-7
IUPAC Name
4-nitrobenzoic acid
Molecular Formula
C7H5NO4
InChI Key
OTLNPYWUJOZPPA-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C=C1)[N+]([O-])=O
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
238.0-244.0?C
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White to cream to yellow
Assay (Silylated GC)
≥98.5%

Description

4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.

Solubility
Very slightly soluble in cold water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text