Thermo Scientific Chemicals

2-Aminobenzamide, 98+%, Thermo Scientific Chemicals

Catalog number: A14756.36
500 g, Each
Thermo Scientific Chemicals

2-Aminobenzamide, 98+%, Thermo Scientific Chemicals

Catalog number: A14756.36
500 g, Each
Quantity
Catalog number: A14756.36
also known as A14756-36
Price (USD)
Quantity
-

Chemical Identifiers

CAS
88-68-6
IUPAC Name
2-aminobenzamide
Molecular Formula
C7H8N2O
InChI Key
PXBFMLJZNCDSMP-UHFFFAOYSA-N
SMILES
NC(=O)C1=CC=CC=C1N
Melting Point (clear melt)
108.0-114.0?C
Identification (FTIR)
Conforms
Appearance (Color)
White brown to grey
Form
Crystals or powder or crystalline powder or flakes
Assay (HPLC)
≥98.0%

Description

2-Aminobenzamide is used as an acetyldehyde scavenger in PET bottles. It is also used as an intermediate for ag chemicals, dyes, pharmaceuticals, blowing agents, and fine organic chemicals. In lubricating oils for supersonic jet engines, inhibits copper and magnesium corrosion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzamide is used as an acetyldehyde scavenger in PET bottles. It is also used as an intermediate for ag chemicals, dyes, pharmaceuticals, blowing agents, and fine organic chemicals. In lubricating oils for supersonic jet engines, inhibits copper and magnesium corrosion.

Solubility
Soluble in hot water and alcohol. Slightly soluble in ether and benzene. Very soluble in ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text