Thermo Scientific Chemicals

O-Methylisourea hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A14773.30
250 g, Each
Thermo Scientific Chemicals

O-Methylisourea hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A14773.30
250 g, Each
Quantity
Catalog number: A14773.30
also known as A14773-30
Price (USD)
Price: 1,601.00
Online price: 1,398.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5329-33-9
IUPAC Name
hydrogen methoxymethanimidamide chloride
Molecular Formula
C2H7ClN2O
InChI Key
MUDVUWOLBJRUGF-UHFFFAOYSA-N
SMILES
[H+].[Cl-].COC(N)=N
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

O-methylisourea hydrochloride is used to prepare 1-chloro-2-methyl-isourea. It is also used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-methylisourea hydrochloride is used to prepare 1-chloro-2-methyl-isourea. It is also used as an organic intermediate.

Solubility
Soluble in organic solvents, water, methanol and ethanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text