Thermo Scientific Chemicals

1,2,3,4-Tetrahydronaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: A14962.0B
1000 g, Each
Thermo Scientific Chemicals

1,2,3,4-Tetrahydronaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: A14962.0B
1000 g, Each
Quantity
Catalog number: A14962.0B
also known as A14962-0B
Price (USD)
Price: 58.10
Online price: 49.65
Your price:
Quantity
-

Chemical Identifiers

CAS
119-64-2
IUPAC Name
1,2,3,4-tetrahydronaphthalene
Molecular Formula
C10H12
InChI Key
CXWXQJXEFPUFDZ-UHFFFAOYSA-N
SMILES
C1CCC2=CC=CC=C2C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Color (APHA)
≤20 (US sourced material)
Assay (GC)
≥96.0% (Non-US sourced material)
Assay from Supplier's CofA
≤96.0% (US sourced material)

Description

1,2,3,4-Tetrahydronaphthalene, is used as an intermediate for organic synthesis, solvent. It is used as a solvent.It is also used for the laboratory synthesis of dry HBr gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydronaphthalene, is used as an intermediate for organic synthesis, solvent. It is used as a solvent.It is also used for the laboratory synthesis of dry HBr gas.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Air and light sensitive. Heat sensitive. May darken on storage. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text