Thermo Scientific Chemicals

2-Amino-6-methylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15187.14
25 g, Each
Thermo Scientific Chemicals

2-Amino-6-methylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15187.14
25 g, Each
Quantity
Catalog number: A15187.14
also known as A15187-14
Price (USD)
Price: 301.00
Online price: 260.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4389-50-8
IUPAC Name
2-amino-6-methylbenzoate
Molecular Formula
C8H8NO2
InChI Key
XHYVBIXKORFHFM-UHFFFAOYSA-M
SMILES
CC1=CC=CC(N)=C1C([O-])=O
Appearance (Color)
Pale cream to cream to pale brown
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%

Description

2-Amino-6-methylbenzoic acid reacts with cyanogen bromide to prepare 2-amino-5-methyl-benzo[d][1,3]oxazin-4-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-methylbenzoic acid reacts with cyanogen bromide to prepare 2-amino-5-methyl-benzo[d][1,3]oxazin-4-one.

Solubility
Soluble in methanol.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text