Thermo Scientific Chemicals

9-Fluorenylmethanol, 99%, Thermo Scientific Chemicals

Catalog number: A15212.22
100 g, Each
Thermo Scientific Chemicals

9-Fluorenylmethanol, 99%, Thermo Scientific Chemicals

Catalog number: A15212.22
100 g, Each
Quantity
Catalog number: A15212.22
also known as A15212-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24324-17-2
IUPAC Name
(9H-fluoren-9-yl)methanol
Molecular Formula
C14H12O
InChI Key
XXSCONYSQQLHTH-UHFFFAOYSA-N
SMILES
OCC1C2=CC=CC=C2C2=CC=CC=C12
Appearance (Color)
White to cream or pale yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
102.0-108.0?C

Description

9-Fluorenylmethanol acts as a N-protecting reagent, which is used in the synthesis of peptide. It is also used in the preparation of deoxynucleoside 9-fluorenemethyl phosphorodithioates. Further, it is used to prepare 9-(fluoromethyl)fluorene. In addition to this, it is involved in the electropolymerization with boron trifluoride diethyl etherate to yield low-potential electrodeposition of semiconducting poly(9-fluorenemethanol) film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Fluorenylmethanol acts as a N-protecting reagent, which is used in the synthesis of peptide. It is also used in the preparation of deoxynucleoside 9-fluorenemethyl phosphorodithioates. Further, it is used to prepare 9-(fluoromethyl)fluorene. In addition to this, it is involved in the electropolymerization with boron trifluoride diethyl etherate to yield low-potential electrodeposition of semiconducting poly(9-fluorenemethanol) film.

Solubility
Soluble in methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text