Thermo Scientific Chemicals

Indole-3-carboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A15330.14
25 g, Each
Thermo Scientific Chemicals

Indole-3-carboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A15330.14
25 g, Each
Quantity
Catalog number: A15330.14
also known as A15330-14
Price (USD)
Price: 81.60
Online price: 69.65
Your price:
Quantity
-

Chemical Identifiers

CAS
487-89-8
IUPAC Name
1H-indole-3-carbaldehyde
Molecular Formula
C9H7NO
InChI Key
OLNJUISKUQQNIM-UHFFFAOYSA-N
SMILES
O=CC1=CNC2=CC=CC=C12
Appearance (Color)
Pale yellow to yellow to pale brown or cream
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.5%
Free acid (titration)
≤0.5%
Melting Point (clear melt)
192.0-198.0?C

Description

Indole-3-carboxaldehyde is a biochemical used to prepare analogs of the indole phytoalexin cyclobrassinin with NR1R2 group. It was also used as the starting material for the synthesis of higher order indoles including isoindolo[2,1-a]indoles, aplysinopsins, and 4-substituted-tetrahydrobenz[cd]indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-carboxaldehyde is a biochemical used to prepare analogs of the indole phytoalexin cyclobrassinin with NR1R2 group. It was also used as the starting material for the synthesis of higher order indoles including isoindolo[2,1-a]indoles, aplysinopsins, and 4-substituted-tetrahydrobenz[cd]indoles.

Solubility
Insoluble in water.

Notes
Air Sensitive. Avoid contact with air/oxygen. Incompatible with reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text