Thermo Scientific Chemicals

1,6-Diiodohexane, 97+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A15368.22
100 g, Each
Thermo Scientific Chemicals

1,6-Diiodohexane, 97+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A15368.22
100 g, Each
Quantity
Catalog number: A15368.22
also known as A15368-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
629-09-4
IUPAC Name
1,6-diiodohexane
Molecular Formula
C6H12I2
InChI Key
QLIMAARBRDAYGQ-UHFFFAOYSA-N
SMILES
ICCCCCCI
Form
Liquid
Refractive Index
1.5840-1.5890 @ 20?C
Appearance (Color)
Clear colorless to yellow to brown
Assay (GC)
≥97.0%
Identification (FTIR)
Conforms

Description

1,6-Diiodohexane is used as a precursor in the preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces. It is also used as a solvent additive and in potential power conversion efficiency of polymer or fullerene bulk heterojunction solar cells. Further, it is employed in the preparation of 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide and 2,4-dihydroxybenzophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Diiodohexane is used as a precursor in the preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces. It is also used as a solvent additive and in potential power conversion efficiency of polymer or fullerene bulk heterojunction solar cells. Further, it is employed in the preparation of 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide and 2,4-dihydroxybenzophenone.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text