Thermo Scientific Chemicals

4-Chlorophenol, 99%, Thermo Scientific Chemicals

Catalog number: A15602.30
250 g, Each
Thermo Scientific Chemicals

4-Chlorophenol, 99%, Thermo Scientific Chemicals

Catalog number: A15602.30
250 g, Each
Quantity
Catalog number: A15602.30
also known as A15602-30
Price (USD)
Price: 59.30
Online price: 50.65
Your price:
Quantity
-

Chemical Identifiers

CAS
106-48-9
IUPAC Name
4-chlorophenol
Molecular Formula
C6H5ClO
InChI Key
WXNZTHHGJRFXKQ-UHFFFAOYSA-N
SMILES
OC1=CC=C(Cl)C=C1
Appearance (Color)
White to cream to pale brown
Form
Crystals or crystalline powder or fused/lumpy solid
Assay (Silylated GC)
≥98.5% (UK-sourced material)
Melting Point
33-46?C
Comment
Material sourced in UK and US

Description

It is used as main raw material of compounding for medicine, dye, plastic and other industries. 4-Chlorophenol was used in visible-light-induced degradation of 4-chlorophenol in aqueous suspension of pure TiO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as main raw material of compounding for medicine, dye, plastic and other industries. 4-Chlorophenol was used in visible-light-induced degradation of 4-chlorophenol in aqueous suspension of pure TiO2.

Solubility
Soluble in water (27g/L).

Notes
Air sensitive. Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text