Thermo Scientific Chemicals

9,10-Diphenylanthracene, 99%, Thermo Scientific Chemicals

Catalog number: A15714.03
1 g, Each
Thermo Scientific Chemicals

9,10-Diphenylanthracene, 99%, Thermo Scientific Chemicals

Catalog number: A15714.03
1 g, Each
Quantity
Catalog number: A15714.03
also known as A15714-03
Price (USD)
Price: 53.10
Online price: 45.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1499-10-1
IUPAC Name
9,10-diphenylanthracene
Molecular Formula
C26H18
InChI Key
FCNCGHJSNVOIKE-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
Form
Crystals or crystalline powder
Assay (HPLC)
>98.5%
Appearance (Color)
Yellow to yellow-green

Description

9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.

Solubility
Soluble in ethanol, chloroform, benzene and carbon disulfide. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text