Thermo Scientific Chemicals

Diethyl succinate, 98%, Thermo Scientific Chemicals

Catalog number: A15777.36
500 g, Each
Thermo Scientific Chemicals

Diethyl succinate, 98%, Thermo Scientific Chemicals

Catalog number: A15777.36
500 g, Each
Quantity
Catalog number: A15777.36
also known as A15777-36
Price (USD)
Price: 42.70
Online price: 36.65
Your price:
Quantity
-

Chemical Identifiers

CAS
123-25-1
IUPAC Name
1,4-diethyl butanedioate
Molecular Formula
C8H14O4
InChI Key
DKMROQRQHGEIOW-UHFFFAOYSA-N
SMILES
CCOC(=O)CCC(=O)OCC
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.4180-1.4220 @ 20?C
Form
Liquid

Description

It is employed as emollients, masking agents, plasticisers and solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as emollients, masking agents, plasticisers and solvents.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text