Thermo Scientific Chemicals

2-Ethyl-4-methylimidazole, 96%, Thermo Scientific Chemicals

Catalog number: A15798.14
25 g, Each
Thermo Scientific Chemicals

2-Ethyl-4-methylimidazole, 96%, Thermo Scientific Chemicals

Catalog number: A15798.14
25 g, Each
Quantity
Catalog number: A15798.14
also known as A15798-14
Price (USD)
Price: 39.60
Online price: 34.65
Your price:
Quantity
-

Chemical Identifiers

CAS
931-36-2
IUPAC Name
2-ethyl-5-methyl-1H-imidazole
Molecular Formula
C6H10N2
InChI Key
ULKLGIFJWFIQFF-UHFFFAOYSA-N
SMILES
CCC1=NC=C(C)N1
Assay (Non-aqueous acid-base Titration)
≥95.0 to ≤105.0%
Form
Lumps or clear liquid or viscous liquid as melt
Assay (GC)
≥95.0%
Identification (FTIR)
Conforms
Refractive Index
1.4990-1.5050 @ 20?C

Description

2-Ethyl-4-methyl-1H-imidazole is a useful synthetic intermediate. It can be used as a building block for active ingredients as well as in epoxy curing. It is used as an intermediate for pharmaceuticals, agrochemicals, dyes, textile auxiliaries, pigments and other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-4-methyl-1H-imidazole is a useful synthetic intermediate. It can be used as a building block for active ingredients as well as in epoxy curing. It is used as an intermediate for pharmaceuticals, agrochemicals, dyes, textile auxiliaries, pigments and other organic chemicals.

Solubility
Soluble in water (<210 g/1000 ml at 20°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text