Thermo Scientific Chemicals

D-Glucurono-6,3-lactone, 99%, Thermo Scientific Chemicals

Catalog number: A15861.14
25 g, Each
Thermo Scientific Chemicals

D-Glucurono-6,3-lactone, 99%, Thermo Scientific Chemicals

Catalog number: A15861.14
25 g, Each
Quantity
Catalog number: A15861.14
also known as A15861-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
32449-92-6
IUPAC Name
(2R)-2-[(2S,3R,4S)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
Molecular Formula
C6H8O6
InChI Key
UYUXSRADSPPKRZ-SKNVOMKLSA-N
SMILES
O[C@@H](C=O)[C@H]1OC(=O)[C@@H](O)[C@H]1O
Acid-base back titration
≥98.5 to ≤101.5%
Solution Test
(5% in water) Clear colorless to pale yellow
Appearance (Color)
White to pale cream
Loss on Drying
≤0.5% (105?C, 3h)
Form
Crystals or powder or crystalline powder

Description

D-Glucurono-6,3-lactone is a glucuronic acid derivative studied for its effectiveness against canine hepatitis. It is utilized in the studies such as starting regent in the synthesis of 2,3,4,-tris(tert.-butyldimethysilyl) glucuronic acid trichloroethylester, required for the preparation of 1-O-acyl glucuronide of the anti-inflammatory drug ML-3000, synthesis of optically active glucopyranoses, synthesis of long-chain alkyl glucofuranosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glucurono-6,3-lactone is a glucuronic acid derivative studied for its effectiveness against canine hepatitis. It is utilized in the studies such as starting regent in the synthesis of 2,3,4,-tris(tert.-butyldimethysilyl) glucuronic acid trichloroethylester, required for the preparation of 1-O-acyl glucuronide of the anti-inflammatory drug ML-3000, synthesis of optically active glucopyranoses, synthesis of long-chain alkyl glucofuranosides.

Solubility
Soluble in Water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text