Thermo Scientific Chemicals

Chlorophyllin, coppered trisodium salt, Thermo Scientific Chemicals

Catalog number: A16155.22
100 g, Each
Thermo Scientific Chemicals

Chlorophyllin, coppered trisodium salt, Thermo Scientific Chemicals

Catalog number: A16155.22
100 g, Each
Quantity
Catalog number: A16155.22
also known as A16155-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
11006-34-1
IUPAC Name
chlorophyllin
Molecular Formula
C34H31CuN4Na3O6
InChI Key
HWDGVJUIHRPKFR-UHFFFAOYNA-I
SMILES
[Na+].[Na+].[Na+].[Cu++].CCC1=C(C)C2=N/C/1=CC1=C(C)C(=C([O-])[O-])C(=N1)C(CC([O-])=O)=C1/N=C(C=C3/[N-]/C(=C2)C(C=C)=C3C)C(C)C1CCC([O-])=O
Appearance (Color)
Dark green to black
Elemental Analysis
Cu: 3.5-6.5%
pH
9.0-11.0 (1% solution in water)
Form
Powder
Loss on Drying
≤10.0% (105°C/constant weight)

Description

Chlorophyllin, coppered trisodium salt is used in the spectrophotometric determination of Ven- lafaxine hydrochloride. A hybrid photoelectrochemical biofuel cell is composed of a TiO 2 film conductive glass electrode sensitized by a copper chlorophyll trisodium salt. Visible light-sensitive gels were prepared by introducing a chromophore (trisodium salt of copper chlorophyllin) to thermosensitive gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorophyllin, coppered trisodium salt is used in the spectrophotometric determination of Ven- lafaxine hydrochloride. A hybrid photoelectrochemical biofuel cell is composed of a TiO 2 film conductive glass electrode sensitized by a copper chlorophyll trisodium salt. Visible light-sensitive gels were prepared by introducing a chromophore (trisodium salt of copper chlorophyllin) to thermosensitive gels.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text