Ammonium fluoride, 96%
Ammonium fluoride, 96%
Ammonium fluoride, 96%
Thermo Scientific Chemicals

Ammonium fluoride, 96%

CAS: 12125-01-8 | FH4N | 37.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A16279.36
also known as A16279-36
Price (EUR)
66,65
Online exclusive
73,50
Save 6,85 (9%)
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
66,65
Online exclusive
73,50
Save 6,85 (9%)
Each
Chemical Identifiers
CAS12125-01-8
IUPAC Nameamine hydrofluoride
Molecular FormulaFH4N
InChI KeyLDDQLRUQCUTJBB-UHFFFAOYSA-N
SMILESN.F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid
Assay (Non-aqueous acid-base Titration)≥95.0 to ≤105.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Ammonium fluoride is used for etching of glass, preservation of food, in printing textile, and as mothproofing agent. It is used in chemical analysis and in brewing. It is used in the synthesis of NaHoF4 and NaEuF4, which are important in solid state lasers as well as photoluminescence. The fluoride ion is used as a base and nucleophile in organic synthesis. Ammonium fluoride providee safe deprotection strategy for the tert-butyldimethylsilyl (TBS) group during RNA synthesis.

Solubility
Freely soluble in water. Slightly soluble in alcohol.

Notes
Hygroscopic and non-cumbustible. On heating, it sublimes and decomposes to hydrogen fluoride and ammonia gas, which can recombine to form ammonium fluoride.
RUO – Research Use Only

General References:

  1. Park, S.; Lim, D.; Seo, Y.; Lee, H. Incorporation of ammonium fluoride into clathrate hydrate lattices and its significance in inhibiting hydrate formation. Chem. Commun. 2015, 51, 8761-8764.
  2. Pesek, J. J.; Matyska, M. T. Ammonium fluoride as a mobile phase additive in aqueous normal phase chromatography. J. Chromatogr. A 2015, 1401, 69-74.