Thermo Scientific Chemicals

3-Fluorobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A16511.22
100 g, Each
Thermo Scientific Chemicals

3-Fluorobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A16511.22
100 g, Each
Quantity
Catalog number: A16511.22
also known as A16511-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
455-38-9
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Melting Point (clear melt)
121.0-127.0?C
Form
Powder or fused solid
Assay (Silylated GC)
≥98.5%

Description

Used in the preparation of regulation of phenolic acid synthesis in Bacillus subtilis. Also used in the bacterial decarboxylation of o-phthalic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of regulation of phenolic acid synthesis in Bacillus subtilis. Also used in the bacterial decarboxylation of o-phthalic acids.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text