Thermo Scientific Chemicals

1-Chloro-3-methyl-2-butene, 95%, stab. with potassium carbonate, Thermo Scientific Chemicals

Catalog number: A16545.18
50 g, Each
Thermo Scientific Chemicals

1-Chloro-3-methyl-2-butene, 95%, stab. with potassium carbonate, Thermo Scientific Chemicals

Catalog number: A16545.18
50 g, Each
Quantity
Catalog number: A16545.18
also known as A16545-18
Price (USD)
Quantity
-

Chemical Identifiers

CAS
503-60-6
IUPAC Name
1-chloro-3-methylbut-2-ene
Molecular Formula
C5H9Cl
InChI Key
JKXQKGNGJVZKFA-UHFFFAOYSA-N
SMILES
CC(C)=CCCl
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4460-1.4520 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥94.0%

Description

1-Chloro-3-methyl-2-butene is used in the synthesis of geraniol. It is also used as an alkylating agent and as a reagent in hyperforin. Further, it is used as an antibiotic to inhibit growth of tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3-methyl-2-butene is used in the synthesis of geraniol. It is also used as an alkylating agent and as a reagent in hyperforin. Further, it is used as an antibiotic to inhibit growth of tumor cells.

Solubility
Miscible with chloroform, acetone, diethyl ether and alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text