Thermo Scientific Chemicals

1,2-Cyclohexanedione dioxime, 97%, Thermo Scientific Chemicals

Catalog number: A16788.06
5 g, Each
Thermo Scientific Chemicals

1,2-Cyclohexanedione dioxime, 97%, Thermo Scientific Chemicals

Catalog number: A16788.06
5 g, Each
Quantity
Catalog number: A16788.06
also known as A16788-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
492-99-9
IUPAC Name
N-[(1E,2E)-2-(hydroxyimino)cyclohexylidene]hydroxylamine
Molecular Formula
C6H10N2O2
InChI Key
CUNNCKOPAWXYDX-KQQUZDAGSA-N
SMILES
O\N=C1/CCCC/C/1=N\O
Form
powder
Elemental Analysis
C: 50.69 ? 2%
Elemental Analysis
N: 19.71 ? 0.7%
Appearance (Color)
White to pale brown

Description

1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.

Solubility
Soluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text