Thermo Scientific Chemicals

N-Benzylethanolamine, 96%, Thermo Scientific Chemicals

Catalog number: A16866.0E
2500 g, Each
Thermo Scientific Chemicals

N-Benzylethanolamine, 96%, Thermo Scientific Chemicals

Catalog number: A16866.0E
2500 g, Each
Quantity
Catalog number: A16866.0E
also known as A16866-0E
Price (USD)
Price: 373.00
Online price: 317.65
Your price:
Quantity
-

Chemical Identifiers

CAS
104-63-2
Assay (GC)
>95.0%
Appearance (Color)
Clear, colorless to pale yellow
Refractive Index
1.5380-1.5450 @ 20?C
Form
Liquid

Description

Has Pharmaceutical applications. N-Benzylethanolamine is used in the synthesis of imidazoles as potent calcitonin (CGRP) antagonists. Also it aids in the preparation of benzofused hydroxamix acids, as useful fragments for the synthesis of histone deacetylase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Has Pharmaceutical applications. N-Benzylethanolamine is used in the synthesis of imidazoles as potent calcitonin (CGRP) antagonists. Also it aids in the preparation of benzofused hydroxamix acids, as useful fragments for the synthesis of histone deacetylase inhibitors.

Solubility
Soluble in water 162 g/L (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents..
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text