Thermo Scientific Chemicals

2-Bromophenylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A16900.22
100 g, Each
Thermo Scientific Chemicals

2-Bromophenylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A16900.22
100 g, Each
Quantity
Catalog number: A16900.22
also known as A16900-22
Price (USD)
Price: 328.00
Online price: 280.65
Your price:
Quantity
-

Chemical Identifiers

CAS
18698-97-0
IUPAC Name
2-(2-bromophenyl)acetate
Molecular Formula
C8H6BrO2
InChI Key
DWXSYDKEWORWBT-UHFFFAOYSA-M
SMILES
[O-]C(=O)CC1=CC=CC=C1Br
Form
Crystalline powder
Assay (HPLC)
>98.0%
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
>98.0 to <102.0%

Description

Employed as a pharmaceutical intermediate. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a pharmaceutical intermediate. It is also used in organic synthesis.

Solubility
Very faint turbidity in methanol.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text