Thermo Scientific Chemicals

3-Heptanone, 98%, Thermo Scientific Chemicals

Catalog number: A16977.AK
250 mL, Each
Thermo Scientific Chemicals

3-Heptanone, 98%, Thermo Scientific Chemicals

Catalog number: A16977.AK
250 mL, Each
Quantity
Catalog number: A16977.AK
also known as A16977-AK
Price (USD)
Price: 90.90
Online price: 79.65
Your price:
Quantity
-

Chemical Identifiers

CAS
106-35-4
IUPAC Name
heptan-3-one
Molecular Formula
C7H14O
InChI Key
NGAZZOYFWWSOGK-UHFFFAOYSA-N
SMILES
CCCCC(=O)CC
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Refractive Index
1.4065-1.4115 @ 20?C
Form
Liquid

Description

Employed as a flavoring agent in melon, nut, and banana flavors. It is also applied as a perfuming agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a flavoring agent in melon, nut, and banana flavors. It is also applied as a perfuming agent in cosmetics industry.

Solubility
Soluble in water at 20°C 4.3g/L.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place. Protect against electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text