Thermo Scientific Chemicals

N,N-Dimethylhexylamine, 99%, Thermo Scientific Chemicals

Catalog number: A17364.06
5 g, Each
Thermo Scientific Chemicals

N,N-Dimethylhexylamine, 99%, Thermo Scientific Chemicals

Catalog number: A17364.06
5 g, Each
Quantity
Catalog number: A17364.06
also known as A17364-06
Price (USD)
Price: 47.00
Online price: 40.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4385-04-0
IUPAC Name
hexyldimethylazanium
Molecular Formula
C8H20N
InChI Key
QMHNQZGXPNCMCO-UHFFFAOYSA-O
SMILES
CCCCCC[NH+](C)C
Identification (FTIR)
Conforms
Refractive Index
1.4120-1.4150 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Form
Liquid

Description

N,N-Dimethylhexylamine is used as an ion-pairing agent to study the poor retention of uridine diphosphate-linked intermediates on reverse phase media. It is also used in the determination of five monophosphate nucleotides in baby foods. It acts as an additive and encapsulated into the nanoparticle with salinomycin (sali), which has selective toxicity to cancer stem cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylhexylamine is used as an ion-pairing agent to study the poor retention of uridine diphosphate-linked intermediates on reverse phase media. It is also used in the determination of five monophosphate nucleotides in baby foods. It acts as an additive and encapsulated into the nanoparticle with salinomycin (sali), which has selective toxicity to cancer stem cells.

Solubility
Miscible with methanol.

Notes
Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text