Thermo Scientific Chemicals

N-tert-Butyl-alpha-phenylnitrone, 97%, Thermo Scientific Chemicals

Catalog number: A17442.06
5 g, Each
Thermo Scientific Chemicals

N-tert-Butyl-alpha-phenylnitrone, 97%, Thermo Scientific Chemicals

Catalog number: A17442.06
5 g, Each
Quantity
Catalog number: A17442.06
also known as A17442-06
Price (USD)
Price: 279.00
Online price: 237.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3376-24-7
IUPAC Name
N-tert-butyl-1-phenylmethanimine oxide
Molecular Formula
C11H15NO
InChI Key
IYSYLWYGCWTJSG-UHFFFAOYSA-N
SMILES
CC(C)(C)[N+]([O-])=CC1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (GC)
≥96.0%
Melting Point (clear melt)
70.0-78.0°C

Description

N-tert-Butyl-a-phenylnitrone is a commonly-used free radical trap. It contains radical scavenging activity and an ability to inhibit Cox-2 (cyclooxygenase-2). N-tert-Butyl-a-phenylnitrone is an antioxidant that has been shown to act as a protective agent in several experimental models of neurodegenerative disorders. N-tert-Butyl-a-phenylnitrone inhibits lipid peroxidation in rat liver microsomes. It also prevents the induction of inducible nitric oxide synthase (iNOS) by reactive oxygen species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-tert-Butyl-a-phenylnitrone is a commonly-used free radical trap. It contains radical scavenging activity and an ability to inhibit Cox-2 (cyclooxygenase-2). N-tert-Butyl-a-phenylnitrone is an antioxidant that has been shown to act as a protective agent in several experimental models of neurodegenerative disorders. N-tert-Butyl-a-phenylnitrone inhibits lipid peroxidation in rat liver microsomes. It also prevents the induction of inducible nitric oxide synthase (iNOS) by reactive oxygen species.

Solubility
Soluble in DMSO (10 mg/ml), chloroform (50 mg/ml), and water (20 mg/ml).

Notes
Store in a cool place. Reactive with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text