Thermo Scientific Chemicals

Nonenylsuccinic anhydride, 96%, Thermo Scientific Chemicals

Catalog number: A17607.30
250 g, Each
Thermo Scientific Chemicals

Nonenylsuccinic anhydride, 96%, Thermo Scientific Chemicals

Catalog number: A17607.30
250 g, Each
Quantity
Catalog number: A17607.30
also known as A17607-30
Price (USD)
Price: 172.00
Online price: 148.65
Your price:
Quantity
-

Chemical Identifiers

CAS
28928-97-4
IUPAC Name
3-[(1E)-non-1-en-1-yl]oxolane-2,5-dione
Molecular Formula
C13H20O3
InChI Key
FGQUIQAGZLBOGL-CMDGGOBGNA-N
SMILES
CCCCCCCC=CC1CC(=O)OC1=O
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Refractive Index
1.4715-1.4775 @20?C
Appearance (Color)
Clear colorless to pale brown
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text