Thermo Scientific Chemicals

4,7-Dimethyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals

Catalog number: A17779.06
5 g, Each
Thermo Scientific Chemicals

4,7-Dimethyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals

Catalog number: A17779.06
5 g, Each
Quantity
Catalog number: A17779.06
also known as A17779-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3248-05-3
IUPAC Name
4,7-dimethyl-1,10-phenanthroline
Molecular Formula
C14H12N2
InChI Key
JIVLDFFWTQYGSR-UHFFFAOYSA-N
SMILES
CC1=C2C=CC3=C(C)C=CN=C3C2=NC=C1
Form
Powder
Assay (GC)
≥97.5%
Appearance (Color)
White to cream to pale brown

Description

4,7-Dimethyl-1,10-phenanthroline acts as an intermediate in synthetic chemistry and dyestuff field. It is also used as an organic bidentate ligand and form complexes with metal for detection. Further, it plays an important role in colorimetric sensors, coordination polymer, ionochromism and luminescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,7-Dimethyl-1,10-phenanthroline acts as an intermediate in synthetic chemistry and dyestuff field. It is also used as an organic bidentate ligand and form complexes with metal for detection. Further, it plays an important role in colorimetric sensors, coordination polymer, ionochromism and luminescence.

Solubility
Soluble in benzene and alcohol. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text