Thermo Scientific Chemicals

Phenyl trans-beta-styryl sulfone, 96%, Thermo Scientific Chemicals

Catalog number: A18556.06
5 g, Each
Thermo Scientific Chemicals

Phenyl trans-beta-styryl sulfone, 96%, Thermo Scientific Chemicals

Catalog number: A18556.06
5 g, Each
Quantity
Catalog number: A18556.06
also known as A18556-06
Price (USD)
Price: 232.00
Online price: 202.65
Your price:
Quantity
-

Chemical Identifiers

CAS
16212-06-9
IUPAC Name
[(1Z)-2-(benzenesulfonyl)ethenyl]benzene
Molecular Formula
C14H12O2S
InChI Key
DNMCCXFLTURVLK-QXMHVHEDSA-N
SMILES
O=S(=O)(C=C/C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder or granules
Assay (GC)
≥95.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
73-76?C

Description

Phenyl trans-beta-styryl sulfone is used as a building block for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl trans-beta-styryl sulfone is used as a building block for proteomics research.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text