Thermo Scientific Chemicals

Methyl 5-bromovalerate, 97%, Thermo Scientific Chemicals

Catalog number: A19072.06
5 g, Each
Thermo Scientific Chemicals

Methyl 5-bromovalerate, 97%, Thermo Scientific Chemicals

Catalog number: A19072.06
5 g, Each
Quantity
Catalog number: A19072.06
also known as A19072-06
Price (USD)
Price: 52.80
Online price: 44.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5454-83-1
IUPAC Name
methyl 5-bromopentanoate
Molecular Formula
C6H11BrO2
InChI Key
RAVVJKCSZXAIQP-UHFFFAOYSA-N
SMILES
COC(=O)CCCCBr
Form
Liquid
Refractive Index
1.4600-1.4650 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%

Description

It is employed as an intermediate in the production of Apixaban .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate in the production of Apixaban .

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text