Thermo Scientific Chemicals

Decanoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A19486.22
100 g, Each
Thermo Scientific Chemicals

Decanoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A19486.22
100 g, Each
Quantity
Catalog number: A19486.22
also known as A19486-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
112-13-0
IUPAC Name
decanoyl chloride
Molecular Formula
C10H19ClO
InChI Key
IPIVAXLHTVNRBS-UHFFFAOYSA-N
SMILES
CCCCCCCCCC(Cl)=O
Appearance (Color)
Clear, colourless to pale yellow
Assay (GC)
> 97.5%
Refractive Index
1.4390 - 1.4430 @20?C
Form
liquid

Description

Decanoyl chloride is used in the synthesis of oligomers composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units. It enhances the activity of proleather from Bacillus sp. during the synthesis of poly(lactic acid) in organic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decanoyl chloride is used in the synthesis of oligomers composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units. It enhances the activity of proleather from Bacillus sp. during the synthesis of poly(lactic acid) in organic solvents.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text