5-Chloro-2-mercaptobenzimidazole, 98%
5-Chloro-2-mercaptobenzimidazole, 98%
5-Chloro-2-mercaptobenzimidazole, 98%
Thermo Scientific Chemicals

5-Chloro-2-mercaptobenzimidazole, 98%

CAS: 25369-78-2 | C7H5ClN2S | 184.641 g/mol
Quantity:
5 g
25 g
Catalog number A19852.06
also known as A19852-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS25369-78-2
IUPAC Name5-chloro-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Molecular FormulaC7H5ClN2S
InChI KeyZZIHEYOZBRPWMB-UHFFFAOYSA-N
SMILESClC1=CC=C2NC(=S)NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale cream
Assay (HPLC)≥97.5%
Melting Point (clear melt)1637.0-974.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only