Collibri™ PS DNA Library Prep Kits for Illumina Systems
Invitrogen™

Collibri™ PS DNA Library Prep Kits for Illumina Systems

The Invitrogen Collibri PS DNA Library Preparation kits are designed for next-generation sequencing (NGS) on Illumina systems. The kit supports whole-genome sequencing (WGS) applications and is suitable for all genomes, including FFPE samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of ReactionsProduct Type
A3861302424 PrepsPS DNA Library Prep Kit with UD Indexes (Set A, 1-24)
A38612024W24 PrepsPS DNA Library Prep Core Kit
A38614096W96 PrepsPS DNA Library Prep Core Kit
A3861202424 PrepsPS DNA Library Prep Kit with CD Indexes
A3861409696 PrepsPS DNA Library Prep Kit with CD Indexes
A4361102424 PrepsPS DNA Library Prep Kit with UD Indexes (Set B, 25-48)
A4361202424 PrepsPS DNA Library Prep Kit with UD Indexes (Set C, 49-72)
A4361302424 PrepsPS DNA Library Prep Kit with UD Indexes (Set D, 73-96)
A3861419696 PrepsPS DNA Library Prep Kit with UD Indexes (Set A-D, 1-96)
Catalog number A38613024
Price (EUR)
1.044,00
Each
No. of Reactions:
24 Preps
Product Type:
PS DNA Library Prep Kit with UD Indexes (Set A, 1-24)
Request bulk or custom format
Price (EUR)
1.044,00
Each
The Collibri PS DNA Library Preparation Kit offers higher confidence in SNP and INDEL calling due to better coverage of challenging genomic regions from a wide range of inputs (1 ng to 1 μg) of physically sheared double-stranded DNA, cfDNA, or amplicons (≤300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 500 ng DNA.

The Collibri PS DNA Library Preparation Kit offers higher confidence in SNP and INDEL calling due to better coverage of challenging genomic regions from a wide range of inputs (1 ng to 1 μg) of physically sheared double-stranded DNA, cfDNA, or amplicons (≤300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 500 ng DNA.

The Collibri PS DNA Library Preparation Kit:
• Includes all necessary components in a single kit* for quick use
• Is empowered on miniaturization liquid handlers to cut on costs
• Provides in-process visual feedback for easier handling
• Is suitable for conventional automation to increase throughput

* The kit consists of enzymatic modification, size selection/clean-up, and indexed adapter-containing modules, except for the Core kit, which enables Collibri combination with desired indexed adapters.

The Collibri PS DNA Library Preparation Kit contains inert dyes that provide visual feedback of preparation progress: the reaction mixes change color in response to the addition of critical components at each step. The kit generates a consistently high yield of library from any amount of input DNA (including the PCR-free version), which is enough for library quantification   and sequencing on Illumina instruments.

Applications
The Collibri PS DNA Library Preparation Kit is recommended for:
• Whole genome sequencing
• Resequencing
• De novo assembly

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionThe product contains modules A39122024, A38587024, A43190024
For Use With (Application)Whole Genome Sequencing, Resequencing, De Novo Assembly
For Use With (Equipment)HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
LibrariesFragment Library
No. of Reactions24 Preps
Product LineCollibri
Product TypePS DNA Library Prep Kit with UD Indexes (Set A, 1-24)
Quantity24 preps
Sample TypeDNA
Shipping ConditionDry Ice
SpeciesAll
Workflow StepLibrary Generation
FormatKit
Sequencing TypeGenome and DNA Sequencing
Unit SizeEach
Contents & Storage
• Collibri PS DNA Library Prep Kit, store at -20°C
• Collibri DNA Library Cleanup Kit, store at 4°C
• Collibri DNA UD Indexes, store at -20°C

Frequently asked questions (FAQs)

Can indexed adaptors or other modules be purchased separately for the Collibri DNA Library Prep kits?

Indexed adapters and other modules in the kit can be purchased separately. Please refer to thermofisher.com/collibriDNA and contact us for more details.

Citations & References (2)

Citations & References
Abstract
Targeted In Situ Protein Diversification and Intra-organelle Validation in Mammalian Cells.
Authors:Erdogan M, Fabritius A, Basquin J, Griesbeck O
Journal:Cell Chem Biol
PubMed ID:32142629
Engineered proteins must be phenotypically selected for function in the appropriate physiological context. Here, we present a versatile approach that allows generating panels of mammalian cells that express diversified heterologous protein libraries in the cytosol or subcellular compartments under stable conditions and in a single-variant-per-cell manner. To this end we adapt ... More
Effects of Prenatal Phthalate Exposure and Childhood Exercise on Maternal Behaviors in Female Rats at Postpartum: A Role of
Authors:Lee YJ, Lin HT, Chaudhary MA, Lee YC, Wang DC
Journal:Int J Mol Sci
PubMed ID:34576011
Both the detrimental effect of prenatal exposure to di-(2-ethylhexyl)-phthalate (DEHP) and the beneficial effects of physical exercise on brain functions have been reported. The oxytocin pathway has been implicated in the onset of maternal behaviors. Epigenetic modification of the oxytocin receptor gene (OXTR) through DNA methylation has been associated with ... More