Orion™ AQUAfast™ AQ3140 COD Colorimeter
Thermo Scientific™

Orion™ AQUAfast™ AQ3140 COD Colorimeter

Designed for the laboratory, this COD colorimeter is a single parameter instrument for determining chemical oxygen demand index in water for markets such as wastewater, food & beverage, aquaculture, and agriculture.
Have Questions?
Catalog NumberTypeIncludesFor Use With (Application)
AQ3140COD ColorimeterColorimeter with field case, batteries and literatureReagents: CODL00, CODL150, CODH00, CODM150, CODHP0, CODH150; Thermoreactor: COD165
Catalog number AQ3140
Price (USD)
882.00
Online exclusive
980.00
Save 98.00
Each
Type:
COD Colorimeter
Includes:
Colorimeter with field case, batteries and literature
For Use With (Application):
Reagents: CODL00, CODL150, CODH00, CODM150, CODHP0, CODH150; Thermoreactor: COD165
Price (USD)
882.00
Online exclusive
980.00
Save 98.00
Each

The Thermo Scientific™ Orion™ AQUAfast™ AQ3140 COD colorimeter is a single parameter instrument for determining chemical oxygen demand in water. This colorimeter is designed to give accurate, reproducible results by offering features such as large digital backlit display, automatic switch-off, user friendly keypad, real time clock and date, calibration mode, storage function, waterproof sample chamber, and many more! The AQUAfast AQ3140 COD colorimeter is used in markets such as wastewater, food & beverage, aquaculture, and agriculture.

The Orion AQUAfast AQ3140 COD colorimeter is ideal for the dedicated COD (chemical oxygen demand) testing of water and wastewater including industrial wastewater and is designed to assist with meeting US EPA testing compliance for COD measurements of wastewater samples. Using two LED light sources at 430 nm and 610nm wavelengths, this colorimeter measures three COD testing ranges of low range 0 to 150mg/L, mid range 0 to 1500mg/L and high range 0 to 15000mg/L. The AQ3140 COD colorimeter is CE certified, IP-67 waterproof and accepts Orion AQUAfast 16mm COD dichromate digestion tubes, available in standard 25 tube packs and new 150 bulk tube packs. The Orion AQUAfast COD165 thermoreactor is also available as a separate accessory for digestion tests.
  • Large digital backlit display
  • Automatic switch-off
  • User-friendly keypad
  • Real-time clock and date
  • Calibration mode
  • Storage function
  • Waterproof sample chamber
  • Angled, ergonomic design for easy bench top or hand-held use
  • 2 year warranty
Specifications
Accuracy3% Full Scale (T = 20 to 25°C)
Backlight OptionYes
Battery Life17 hours operating time or 5,000 measurements in continuous mode when display backlight is off
Battery Type4 x AAA
CalibrationUser and factory calibration functions
Certifications/ComplianceCE
Cuvette Details16 mm COD Digestion Tubes
Data Storage16 with date/time stamp
Dimensions (L x W x H)155 x 75 x 35 mm
For Use With (Application)Reagents: CODL00, CODL150, CODH00, CODM150, CODHP0, CODH150; Thermoreactor: COD165
IP RatingIP67
IncludesColorimeter with field case, batteries and literature
PackagingDigestion tubes
RangeLow Range: 0 to 150 mg/L O2; Mid Range: 0 to 1500 mg/L O2; High Range: 0 to 15000 mg/L O2
Test MethodDedicated Chlorine Colorimeter
Warranty2 Years
Wavelength Accuracy± 1 nm
Weight (Metric)260 g (With batteries)
DisplayBacklit LCD
Light Source2 x LED
Product LineAQUAfast
TypeCOD Colorimeter
Wavelength Range430, 610 nm
Unit SizeEach