Thermo Scientific Chemicals

1-Acetylpiperazine, 98%, Thermo Scientific Chemicals

Catalog number: B20200.14
25 g, Each
Thermo Scientific Chemicals

1-Acetylpiperazine, 98%, Thermo Scientific Chemicals

Catalog number: B20200.14
25 g, Each
Quantity
Catalog number: B20200.14
also known as B20200-14
Price (USD)
Price: 135.00
Online price: 115.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13889-98-0
IUPAC Name
4-acetylpiperazin-1-ium
Molecular Formula
C6H13N2O
InChI Key
PKDPUENCROCRCH-UHFFFAOYSA-O
SMILES
CC(=O)N1CC[NH2+]CC1
Form
Crystals or powder or crystalline powder or fused solid or liquid as melt
Assay (GC)
≥98.5%
Appearance (Color)
White to cream to yellow to green
Melting Point (clear melt)
28.0-35.0?C

Description

1-Acetylpiperazine may be used in the synthesis of series of 7-alkoxyl substituted indolizinoquinoline-5,12-dione derivatives and 2-substituted-N-(naphth-1-ylmethyl)-pyrimidin-4-amines and 2-substituted-N-benzhydrylpyrimidin-4-amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Acetylpiperazine may be used in the synthesis of series of 7-alkoxyl substituted indolizinoquinoline-5,12-dione derivatives and 2-substituted-N-(naphth-1-ylmethyl)-pyrimidin-4-amines and 2-substituted-N-benzhydrylpyrimidin-4-amines.

Solubility
Soluble in water 210 g/L (20°C).

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text