Thermo Scientific Chemicals

4-(4-Morpholinyl)aniline, 98+%, Thermo Scientific Chemicals

Catalog number: B20292.14
25 g, Each
Thermo Scientific Chemicals

4-(4-Morpholinyl)aniline, 98+%, Thermo Scientific Chemicals

Catalog number: B20292.14
25 g, Each
Quantity
Catalog number: B20292.14
also known as B20292-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2524-67-6
IUPAC Name
4-(morpholin-4-yl)aniline
Molecular Formula
C10H14N2O
InChI Key
PHNDZBFLOPIMSM-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=C1)N1CCOCC1
Appearance (Color)
White to cream or pink to brown
Melting Point (clear melt)
128.5-133.5?C
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%

Description

4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Morpholinyl)aniline is used in the preparation of central nervous system (CNS) active agents, also used in the color developing process in photographs.

Solubility
Soluble in chloroform, ethyl acetate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, acid hydrides, acid chlorides, chloroformates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text