Thermo Scientific Chemicals

Sodium 1-undecanesulfonate, 98+% (dry wt.), water <2%, Thermo Scientific Chemicals

Catalog number: B20332.22
100 g, Each
Thermo Scientific Chemicals

Sodium 1-undecanesulfonate, 98+% (dry wt.), water <2%, Thermo Scientific Chemicals

Catalog number: B20332.22
100 g, Each
Quantity
Catalog number: B20332.22
also known as B20332-22
Price (USD)
Price: 619.00
Online price: 550.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5838-34-6
Form
Crystals/crystalline powder
Sulfated Ash
26.49-28.49% (Theoretical 27.49%)
Appearance (Color)
White
Loss on Drying
<2.0%
Assay (ex Sulphated Ash)
>98.0% (dry wt. basis)

Description

It can be used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as catalyst in pharmaceutical research.

Solubility
Solubility in water is almost transparent.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text