Thermo Scientific Chemicals

Hyodeoxycholic acid, 96%, Thermo Scientific Chemicals

Catalog number: B20506.14
25g, Each
Thermo Scientific Chemicals

Hyodeoxycholic acid, 96%, Thermo Scientific Chemicals

Catalog number: B20506.14
25g, Each
Quantity
Catalog number: B20506.14
also known as B20506-14
Price (USD)

Chemical Identifiers

CAS
83-49-8
IUPAC Name
(4R)-4-[(1R,3aS,3bS,5S,5aR,7R,9aR,9bS,11aR)-5,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Molecular Formula
C24H40O4
InChI Key
DGABKXLVXPYZII-SIBKNCMHSA-N
SMILES
[H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])C[C@H](O)[C@]2([H])C[C@H](O)CC[C@]12C
Appearance (Color)
White
Assay from Supplier's CofA
≥95.0% (U.S. specification)
Optical Rotation
7.8 ± 0.1° (C=2 in ethanol, non-U.S. specification)
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0% (non-U.S. specification)
Melting Point (clear melt)
193.0-202.0°C

Description

It is the main acid constituent of hog bile, and for this reason it was used industrially as precursor for steroid synthesis before total synthesis became practical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the main acid constituent of hog bile, and for this reason it was used industrially as precursor for steroid synthesis before total synthesis became practical.

Solubility
Solubility in methanol, very faint turbidity. Slightly soluble in alcohol, acetone, ether and very slightly soluble in chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text