Thermo Scientific Chemicals

3-Decyn-1-ol, 97%, Thermo Scientific Chemicals

Catalog number: B20837.14
25 g, Each
Thermo Scientific Chemicals

3-Decyn-1-ol, 97%, Thermo Scientific Chemicals

Catalog number: B20837.14
25 g, Each
Quantity
Catalog number: B20837.14
also known as B20837-14
Price (USD)
Price: 426.00
Online price: 412.65
Your price:
Quantity
-

Chemical Identifiers

CAS
51721-39-2
IUPAC Name
dec-3-yn-1-ol
Molecular Formula
C10H18O
InChI Key
YGEQBZUDPQQIFI-UHFFFAOYSA-N
SMILES
CCCCCCC#CCCO
Form
Liquid
Refractive Index
1.4555-1.4605 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%

Description

3-Decyn-1-ol may be used in the following studies, as a starting material in the synthesis of 3?-deoxyribolactones, as a starting material in the synthesis of (Z)-3-decen-1-ol by hydrogenation over P-2 nickel and as a reagent in the synthesis of methyl 9-decynoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Decyn-1-ol may be used in the following studies, as a starting material in the synthesis of 3′-deoxyribolactones, as a starting material in the synthesis of (Z)-3-decen-1-ol by hydrogenation over P-2 nickel and as a reagent in the synthesis of methyl 9-decynoate.

Solubility
Sparingly soluble in water (0.21 g/L) (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text