Thermo Scientific Chemicals

1-Phenyl-1-pentyne, 98%, Thermo Scientific Chemicals

Catalog number: B20857.14
25 g, Each
Thermo Scientific Chemicals

1-Phenyl-1-pentyne, 98%, Thermo Scientific Chemicals

Catalog number: B20857.14
25 g, Each
Quantity
Catalog number: B20857.14
also known as B20857-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4250-81-1
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.5400-1.5440 @ 20?C
Assay (GC)
≥97.5%

Description

It finds it application in the highly regioselective synthesis of polysubstituted naphthalene derivatives through gallium trichloride catalyzed alkyne-aldehyde coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application in the highly regioselective synthesis of polysubstituted naphthalene derivatives through gallium trichloride catalyzed alkyne-aldehyde coupling.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text