Thermo Scientific Chemicals

(S)-(-)-1-Phenylethanol, 97+%, Thermo Scientific Chemicals

Catalog number: B21188.06
5 g, Each
Thermo Scientific Chemicals

(S)-(-)-1-Phenylethanol, 97+%, Thermo Scientific Chemicals

Catalog number: B21188.06
5 g, Each
Quantity
Catalog number: B21188.06
also known as B21188-06
Price (USD)
Price: 155.00
Online price: 131.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1445-91-6
IUPAC Name
(1S)-1-phenylethan-1-ol
Molecular Formula
C8H10O
InChI Key
WAPNOHKVXSQRPX-ZETCQYMHSA-N
SMILES
C[C@H](O)C1=CC=CC=C1
Form
Liquid
Assay (GC)
≥97.0%
Optical Rotation
-45.0 ± 1.0° (c=5 in methanol)
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms

Description

It finds its application in the synthesis of optically active products. Used for the synthesis of various pharmaceutical compounds. It can also be used as a chiral derivatizing agent for determining the absolute configuration of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the synthesis of optically active products. Used for the synthesis of various pharmaceutical compounds. It can also be used as a chiral derivatizing agent for determining the absolute configuration of secondary alcohols.

Solubility
Soluble in water at 20°C 20g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text