1,3,5-Triphenylbenzene, 99+%
1,3,5-Triphenylbenzene, 99+%
1,3,5-Triphenylbenzene, 99+%
Thermo Scientific Chemicals

1,3,5-Triphenylbenzene, 99+%

CAS: 612-71-5 | C24H18 | 306.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number B21353.30
also known as B21353-30
Price (USD)
230.00
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
230.00
Each
Chemical Identifiers
CAS612-71-5
IUPAC Name3,5-diphenyl-1,1'-biphenyl
Molecular FormulaC24H18
InChI KeySXWIAEOZZQADEY-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥99.0%
Melting Point (clear melt)171.0-177.0?C
1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.

Solubility
Solubility in toluene is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Y. C. Lin and D. E. Williams. 1,3,5-Triphenylbenzene.Acta Cryst.1975B31318-320.
  2. Sakan Sirilaksanapong, et al. 1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.Chem. Commun., 201248(2), 293-295.