Thermo Scientific Chemicals

n-Dodecylbenzene, 97%, Thermo Scientific Chemicals

Catalog number: B21362.22
100 g, Each
Thermo Scientific Chemicals

n-Dodecylbenzene, 97%, Thermo Scientific Chemicals

Catalog number: B21362.22
100 g, Each
Quantity
Catalog number: B21362.22
also known as B21362-22
Price (USD)
Price: 310.00
Online price: 268.65
Your price:
Quantity
-

Chemical Identifiers

CAS
123-01-3
IUPAC Name
dodecylbenzene
Molecular Formula
C18H30
InChI Key
KWKXNDCHNDYVRT-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4785-1.4835 @ 20?C

Description

n-Dodecylbenzene, is a precursor to sodium dodecylbenzenesulfonate, a surfactant that is a key ingredient of household laundry detergents, such as Tide. It is a widely used cleaner, dyeing assistant in textile industrial, degrease agent in electroplate, leather, de- inking agent in papering, rubber and plastic, break emulsion agent in oil industry, artesian well foaming agent in oil well.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Dodecylbenzene, is a precursor to sodium dodecylbenzenesulfonate, a surfactant that is a key ingredient of household laundry detergents, such as Tide. It is a widely used cleaner, dyeing assistant in textile industrial, degrease agent in electroplate, leather, de- inking agent in papering, rubber and plastic, break emulsion agent in oil industry, artesian well foaming agent in oil well.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, nitric acid.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text