Thermo Scientific Chemicals

Dimethyl 5-hydroxyisophthalate, 98%, Thermo Scientific Chemicals

Catalog number: B21565.09
10 g, Each
Thermo Scientific Chemicals

Dimethyl 5-hydroxyisophthalate, 98%, Thermo Scientific Chemicals

Catalog number: B21565.09
10 g, Each
Quantity
Catalog number: B21565.09
also known as B21565-09
Price (USD)
Price: 35.00
Online price: 29.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13036-02-7
IUPAC Name
1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate
Molecular Formula
C10H10O5
InChI Key
DOSDTCPDBPRFHQ-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(=CC(O)=C1)C(=O)OC
Melting Point (clear melt)
160.0-167.0?C
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Assay (HPLC)
≥97.5%

Description

Dimethyl 5-Hydroxyisophthalate is a useful synthetic intermediate in the synthesis of N,N’-Bis(2,3-dihydroxypropyl)-5-[(N-(2-hydroxyethyl)carbamoyl)methoxy]-2,4,6-triiodoisophthalamide (B426310); a compound related to Ioversol( I737000) which is a nonionic, low osmolality, radiographic contrast agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 5-Hydroxyisophthalate is a useful synthetic intermediate in the synthesis of N,N’-Bis(2,3-dihydroxypropyl)-5-[(N-(2-hydroxyethyl)carbamoyl)methoxy]-2,4,6-triiodoisophthalamide (B426310); a compound related to Ioversol( I737000) which is a nonionic, low osmolality, radiographic contrast agent.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text