Thermo Scientific Chemicals

2-Chloro-5-methylthiophene, 97%, Thermo Scientific Chemicals

Catalog number: B21657.06
5 g, Each
Thermo Scientific Chemicals

2-Chloro-5-methylthiophene, 97%, Thermo Scientific Chemicals

Catalog number: B21657.06
5 g, Each
Quantity
Catalog number: B21657.06
also known as B21657-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
17249-82-0
IUPAC Name
2-chloro-5-methylthiophene
Molecular Formula
C5H5ClS
InChI Key
JSMMZMYGEVUURX-UHFFFAOYSA-N
SMILES
CC1=CC=C(Cl)S1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5340-1.5390 @ 20?C

Description

It is used in the synthesis of dithienylcyclopentene optical molecular switches. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of dithienylcyclopentene optical molecular switches. It is also used as an active pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text