Thermo Scientific Chemicals

1-Methylcyclohexanecarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: B21713.18
50 g, Each
Thermo Scientific Chemicals

1-Methylcyclohexanecarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: B21713.18
50 g, Each
Quantity
Catalog number: B21713.18
also known as B21713-18
Price (USD)
Price: 287.00
Online price: 245.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1123-25-7
IUPAC Name
1-methylcyclohexane-1-carboxylate
Molecular Formula
C8H13O2
InChI Key
REHQLKUNRPCYEW-UHFFFAOYSA-M
SMILES
CC1(CCCCC1)C([O-])=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or fused solid
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (Silylated GC)
≥98.5%
Melting Point (clear melt)
35.0-41.0?C

Description

1-Methyl-1-cyclohexanecarboxylic acid was used as internal standard during the determination of valproic acid metabolites. Also been used as an anticonvulsant drug and causes maturation of murine neuroblastoma cells in vitro. Used in the synthesis of p38 map kinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text