Thermo Scientific Chemicals

2-Aminobenzenesulfonamide, 98%, Thermo Scientific Chemicals

Catalog number: B21912.22
100 g, Each
Thermo Scientific Chemicals

2-Aminobenzenesulfonamide, 98%, Thermo Scientific Chemicals

Catalog number: B21912.22
100 g, Each
Quantity
Catalog number: B21912.22
also known as B21912-22
Price (USD)
Price: 873.00
Online price: 762.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3306-62-5
IUPAC Name
2-aminobenzene-1-sulfonamide
Molecular Formula
C6H8N2O2S
InChI Key
YAZSBRQTAHVVGE-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1S(N)(=O)=O
Appearance (Color)
White to cream to pale brown
Assay (Non-aqueous acid-base Titration)
≥97.5% to ≤102.5%
Identification (FTIR)
Conforms
Assay (HPLC)
≥97.5%
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text