Thermo Scientific Chemicals

1-(Trifluoromethyl)vinyl acetate, 97%, Thermo Scientific Chemicals

Catalog number: B22133.18
50 g, Each
Thermo Scientific Chemicals

1-(Trifluoromethyl)vinyl acetate, 97%, Thermo Scientific Chemicals

Catalog number: B22133.18
50 g, Each
Quantity
Catalog number: B22133.18
also known as B22133-18
Price (USD)
Quantity
-

Description

1-(Trifluoromethyl)vinyl acetate is used as a chemical, organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Trifluoromethyl)vinyl acetate is used as a chemical, organic Intermediates.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, heat, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text